missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR123 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575823
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. G protein coupled receptors translate extracellular signals into intracellular signals (G protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. GPR123 (G protein-coupled receptor 123) is a 1,279 amino acid multi-pass membrane protein belonging to the G-protein coupled receptor 2 family and LN-TM7 subfamily. Existing as two alternatively spliced isoforms, GPR123 functions as an orphan receptor that is expressed in adult and fetal brain, and in adult spinal cord. The gene encoding GPR123 maps to human chromosome 10q26.3 and mouse chromosome 7 F4.
Specifications
| GPR123 | |
| Polyclonal | |
| Unconjugated | |
| ADGRA1 | |
| 2900059M17Rik; Adgra1; Adhesion G protein-coupled receptor A1; D7Ertd680e; G protein-coupled receptor 123; GPR123; G-protein coupled receptor 123; KIAA1828; probable G-protein coupled receptor 123 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 309097, 52389, 84435 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86SQ6, Q8C4G9 | |
| ADGRA1 | |
| Synthetic peptide corresponding to residues in Human GPR123. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction