missing translation for 'onlineSavingsMsg'
Learn More

GPR120 modulator 2, MedChemExpress

Product Code. 16480528
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480528 5 mg 5mg
16438068 10 mg 10mg
16423987 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16480528 Supplier Medchem Express Supplier No. HY50172

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GPR120 modulator 2 is a G protein coupled receptor 120 (GPR120) modulator extracted from patent US8394841B2, compound example F13. GPR120 modulator 2 can be used for the research of diseases associated with abnormal or deregulated GPR120, such as diabetes.

GPR120 modulator 2 is a G protein coupled receptor 120 (GPR120) modulator extracted from patent US8394841B2, compound example F13. GPR120 modulator 2 can be used for the research of diseases associated with abnormal or deregulated GPR120, such as diabetes.

Specifications

Chemical Name or Material GPR120 modulator 2
CAS 1050506-87-0
Color White
Purity Grade Notes Research
Molecular Formula C20H18ClNO3S
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (257.81 mM)
SMILES O=C(O)CCC1=CC=C(OCC2=NC(C3=CC=CC=C3Cl)=CS2)C=C1C
Molecular Weight (g/mol) 387.88
Formula Weight 387.88
Percent Purity 98.06%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.