missing translation for 'onlineSavingsMsg'
Learn More

GPR120 Agonist 2, MedChemExpress

Product Code. p-200096954
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490737 50 mg 50mg
16477147 5 mg 5mg
16455316 100 mg 100mg
16444428 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16490737 Supplier Medchem Express Supplier No. HY111353

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GPR120 Agonist 2 is a GPR120 agonist extracted from patent US 20110313003 A1, example 209.

GPR120 Agonist 2 is a GPR120 agonist extracted from patent US 20110313003 A1, example 209.

Specifications

Chemical Name or Material GPR120 Agonist 2
CAS 1234844-11-1
Color White
Purity Grade Notes Research
Molecular Formula C22H25ClO4
Quantity 50 mg
Solubility Information DMSO : ≥ 50 mg/mL (128.57 mM)
SMILES O=C(O)CCC1=CC=C(OCC2=C(OC(C)(C)C3)C3=CC(Cl)=C2)C(C)=C1C
Molecular Weight (g/mol) 388.88
Formula Weight 388.88
Percent Purity 99.07%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.