missing translation for 'onlineSavingsMsg'
Learn More

GPR12 Antibody - BSA Free, Novus Biologicals™

Product Code. 18420994 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420994 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420994 Supplier Novus Biologicals Supplier No. NBP102318

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GPR12 Polyclonal antibody specifically detects GPR12 in Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Hamster, Primate, Monkey samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR12
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 10 μg/mL, Immunohistochemistry-Paraffin 10 μg/mL
Formulation PBS
Gene Accession No. P47775
Gene Alias FLJ18149, FLJ97704, G protein-coupled receptor 12, GPCR12, GPCR21, G-protein coupled receptor 12, MGC138349, MGC138351
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from 1st extracellular domain of human GPR12.
Purification Method Immunogen affinity purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2835
Target Species Human, Mouse, Rat, Pig, Bovine
Content And Storage Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.