missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR119 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577449
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
GPCR GPR119 is a member of the G protein coupled receptor (GPCR) family. Members of this protein family contain seven putative transmembrane domains and may mediate signaling processes to the interior of the cell via activation of heterotrimeric G proteins.
Specifications
| GPR119 | |
| Polyclonal | |
| Unconjugated | |
| Gpr119 | |
| Glucose Dependent Insulinotropic Receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 236781, 302813 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 0.85 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| Q7TQN8, Q7TQP3 | |
| Gpr119 | |
| Peptide CDMLK IASVHSQHIR, corresponding to amino acid residues 186-200 of rat Glucose-dependent Insulin tropic Receptor | |
| 50 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction