missing translation for 'onlineSavingsMsg'
Learn More

GPR119 Antibody, DyLight 550, Novus Biologicals™

Product Code. 18321467 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mL
Unit Size:
0.05mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18321467 0.05 mL 0.05mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18321467 Supplier Novus Biologicals Supplier No. NLS548R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GPR119 Polyclonal specifically detects GPR119 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR119
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Dilution Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:100
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q8TDV5
Gene Alias G protein-coupled receptor 119, glucose-dependent insulinotropic receptor, GPCR2, G-protein coupled receptor 119, G-protein coupled receptor 2, hGPCR2, MGC119957
Gene Symbols GPR119
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human GPR119 protein (within residues 150-250). [Swiss-Prot Q8TDV5]
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 139760
Test Specificity Human and mouse. Immunogen sequence has 88% identity to rat.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.