missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR119 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£196.00 - £363.00
Specifications
| Antigen | GPR119 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:100 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18327945
|
Novus Biologicals
NLS548SS |
0.025 mL |
£196.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18345664
|
Novus Biologicals
NLS548 |
0.05 mL |
£363.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GPR119 Polyclonal specifically detects GPR119 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| GPR119 | |
| Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:100 | |
| Polyclonal | |
| Rabbit | |
| GPCR | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| G protein-coupled receptor 119, glucose-dependent insulinotropic receptor, GPCR2, G-protein coupled receptor 119, G-protein coupled receptor 2, hGPCR2, MGC119957 | |
| GPR119 | |
| IgG | |
| Affinity Purified | |
| Human and mouse. Immunogen sequence has 88% identity to rat. |
| 1 mg/mL | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Q8TDV5, Q7TQP3 | |
| 139760 | |
| A synthetic peptide made to an internal region of the human GPR119 protein (within residues 150-250). [Swiss-Prot Q8TDV5] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title