missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR116 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £375.00
Specifications
| Antigen | GPR116 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18306136
|
Novus Biologicals
NBP3-03588-20UL |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18358764
|
Novus Biologicals
NBP3-03588-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GPR116 Polyclonal specifically detects GPR116 in Mouse, Rat samples. It is validated for Western Blot.Specifications
| GPR116 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| FLJ90640, G protein-coupled receptor 116, Ig-Hepta homolog, KIAA0758DKFZp564O1923, KPG_001, probable G-protein coupled receptor 116 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 22-280 of human GPR116 (NP_056049.4). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| GPCR | |
| PBS with 50% glycerol, pH7.3. | |
| 221395 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title