Learn More
GPR116 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14047-1-AP-20UL
 View more versions of this product
Product Code. 16852514
  • £135.00 / 20µL
Description
G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. G protein coupled receptors translate extracellular signals into intracellular signals (G protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. GPR116 (G protein-coupled receptor 116) is a 1,346 amino acid multi-pass membrane protein that contains one SEA domain, one GPS domain and three Ig-like domains and belongs to the GPR family. Existing as a dilsulfide-liked homodimer at the cell surface, GPR116 exists as multiple alternatively spliced isoforms and is thought to play a role in regulating and maintaining proper acid-base balance throughout the cell.
Specifications
Specifications
GPR116
Polyclonal
Unconjugated
ADGRF5
G protein coupled receptor 116, GPR116, KIAA0758, KPG_001
Rabbit
Antigen Affinity Chromatography
RUO
221395, 224792, 245977
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
G5E8Q8, Q8IZF2, Q9WVT0
ADGRF5
GPR116 Fusion Protein Ag5214
20 μL
Primary
Human, Rat, Mouse