missing translation for 'onlineSavingsMsg'
Learn More

GPR115 Polyclonal Antibody, Invitrogen™

Product Code. 11586172
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11586172 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11586172 Supplier Invitrogen Supplier No. PA120400

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPCR GPR115 is a member of this family (subfamily Orphan-B). ESTs have been isolated from breast, heart, and placenta libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR115
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene ADGRF4
Gene Accession No. Q8IZF3
Gene Alias 4632435A09Rik; ADGRF4; Adhesion G protein-coupled receptor F4; AV239010; FLJ38076; G protein-coupled receptor 115; GPR115; G-protein coupled receptor 115; G-protein coupled receptor PGR18; PGR18; probable G-protein coupled receptor 115; RGD1565216; seven transmembrane helix receptor
Gene Symbols ADGRF4
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from C-terminus of human GPR115.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221393
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.