missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR115 Monoclonal Antibody (527018)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561555
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPCR GPR115 is a member of this family (subfamily Orphan-B). ESTs have been isolated from breast, heart, and placenta libraries.
Specifications
| GPR115 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8IZF3 | |
| ADGRF4 | |
| Chinese hamster ovary cell line CHO-derived recombinant human GPR115, Ser22-Ala347. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| 527018 | |
| Unconjugated | |
| ADGRF4 | |
| 4632435A09Rik; ADGRF4; Adhesion G protein-coupled receptor F4; AV239010; FLJ38076; G protein-coupled receptor 115; GPR115; G-protein coupled receptor 115; G-protein coupled receptor PGR18; PGR18; probable G-protein coupled receptor 115; RGD1565216; seven transmembrane helix receptor | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 221393 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto