missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR115 Antibody (527018) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB54371JF669
This item is not returnable.
View return policy
Description
GPR115 Monoclonal antibody specifically detects GPR115 in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| GPR115 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| FLJ38076, G protein-coupled receptor 115, G-protein coupled receptor PGR18, PGR18G-protein coupled receptor 115, probable G-protein coupled receptor 115, seven transmembrane helix receptor | |
| Chinese hamster ovary cell line CHO-derived recombinant human GPR115, Ser22-Ala347, Accession # Q8IZF3 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Western Blot, CyTOF | |
| 527018 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| GPCR | |
| 221393 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction