missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR113 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06237-50ul
This item is not returnable.
View return policy
Description
GPR113 Polyclonal antibody specifically detects GPR113 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| GPR113 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| FLJ16767, G protein-coupled receptor 113, G-protein coupled receptor 113, G-protein coupled receptor PGR23, hGPCR37, PGR23G protein-coupled receptor PGR23, probable G-protein coupled receptor 113, seven transmembrane helix receptor | |
| Produced in rabbits immunized with E. coli-derived Human GPR113 fragment. | |
| 50 μg | |
| GPCR | |
| 165082 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction