missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR111 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£349.00
Spécification
| Antigen | GPR111 |
|---|---|
| Concentration | 0.33 mg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16883485
|
Proteintech
24965-1-AP-20UL |
20 μL |
N/A
|
N/A | |||||
|
16873485
|
Proteintech
24965-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| GPR111 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q8IZF7 | |
| 222611 | |
| GPR111 Fusion Protein Ag21705 | |
| Primary | |
| -20°C | |
| ADGRF2 |
| 0.33 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| G protein coupled receptor 111, GPR111, hGPCR35, PGR20 | |
| ADGRF2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title