missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR111 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA547845
This item is not returnable.
View return policy
Description
In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) GPR30, rhGPR56, rhGPR114, rhGPR115, rhGPR124, and rhGPR125 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
Specifications
| GPR111 | |
| Polyclonal | |
| Unconjugated | |
| ADGRF2 | |
| ADGRF2; adhesion G protein-coupled receptor F2; adhesion G-protein coupled receptor F2; G protein-coupled receptor 111; GPR111; G-protein coupled receptor 111; G-protein coupled receptor PGR20; hGPCR35; PGR20; probable G-protein coupled receptor 111 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 222611 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q8IZF7 | |
| ADGRF2 | |
| CHO-derived recombinant human GPR111 Cys19-Ser375. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction