missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR111 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116180
This item is not returnable.
View return policy
Description
GPR111 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| GPR111 | |
| Polyclonal | |
| Unconjugated | |
| ADGRF2 | |
| ADGRF2; adhesion G protein-coupled receptor F2; adhesion G-protein coupled receptor F2; G protein-coupled receptor 111; GPR111; G-protein coupled receptor 111; G-protein coupled receptor PGR20; hGPCR35; PGR20; probable G-protein coupled receptor 111 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 222611 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.25 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8IZF7 | |
| ADGRF2 | |
| KLH conjugated synthetic peptide between 366-393 amino acids from the Central region of human GP111. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction