missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR110 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102059
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GPR110.
G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. G protein coupled receptors translate extracellular signals into intracellular signals (G protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. GPR110 (G protein-coupled receptor 110), also known as PGR19, is a 911 amino acid protein that belongs to the G-protein coupled receptor 2 family and LN-TM7 subfamily. Characterized as an adhesion GPCR, GPR110 is a multipass membrane-bound protein with a long amino-terminus that contains multiple domains. One of these domains is the GPCR proteolytic site (GPS), which is essential for proteolytic cleavage of the amino-terminus and for cell surface expression.
Specifications
| GPR110 | |
| Polyclonal | |
| Unconjugated | |
| ADGRF1 | |
| 5031409J19Rik; Adgrf1; adhesion G protein-coupled receptor F1; adhesion G-protein coupled receptor F1; G protein-coupled receptor 110; G protein-coupled receptor KPG_012; G protein-coupled receptor PGR19; Gpr110; G-protein coupled receptor 110; G-protein coupled receptor KPG_012; hGPCR36; internal gene name of KIRIN laboratory:M5; KPG_009; KPG_012; PGR19; probable G-protein coupled receptor 110; seven transmembrane helix receptor | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 266977 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5T601 | |
| ADGRF1 | |
| A synthesized peptide derived from human ADGRF1(Accession Q5T601), corresponding to amino acid residues G838-H888. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur