missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR109B Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533605
This item is not returnable.
View return policy
Description
BLAST analysis of the peptide immunogen showed 100% homology with human protein HCAR2. Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Panda (89%).
Receptor for 3-OH-octanoid acid mediates a negative feedback regulation of adipocyte lipolysis to counteract prolipolytic influences under conditions of physiological or pathological increases in beta-oxidation rates. Acts as a low affinity receptor for nicotinic acid. This pharmacological effect requires nicotinic acid doses that are much higher than those provided by a normal diet. [UniProt].
Specifications
| GPR109B | |
| Polyclonal | |
| Unconjugated | |
| HCAR3 | |
| G protein-coupled receptor 109B; GPR109B; G-protein coupled receptor 109B; G-protein coupled receptor HM74; G-protein coupled receptor HM74B; GTP-binding protein; HCA3; HCAR3; HM74; HM74B; hydroxycarboxylic acid receptor 3; hydroxy-carboxylic acid receptor 3; Niacin receptor 2; NIACR2; Nicotinic acid receptor 2; PUMAG; Puma-g; putative chemokine receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8843 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P49019 | |
| HCAR3 | |
| Synthetic 18 amino acid peptide from N-terminal extracellular domain of human GPR109B / HM74. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction