missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR109B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106832
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GPR109.
Receptor for 3-OH-octanoid acid mediates a negative feedback regulation of adipocyte lipolysis to counteract prolipolytic influences under conditions of physiological or pathological increases in beta-oxidation rates. Acts as a low affinity receptor for nicotinic acid. This pharmacological effect requires nicotinic acid doses that are much higher than those provided by a normal diet. [UniProt].
Specifications
| GPR109B | |
| Polyclonal | |
| Unconjugated | |
| HCAR3 | |
| G protein-coupled receptor 109B; GPR109B; G-protein coupled receptor 109B; G-protein coupled receptor HM74; G-protein coupled receptor HM74B; GTP-binding protein; HCA3; HCAR3; HM74; HM74B; hydroxycarboxylic acid receptor 3; hydroxy-carboxylic acid receptor 3; Niacin receptor 2; NIACR2; Nicotinic acid receptor 2; PUMAG; Puma-g; putative chemokine receptor | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 369162, 8843 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P49019 | |
| HCAR3 | |
| A synthesized peptide derived from human HCAR3(Accession P49019), corresponding to amino acid residues P302-P352. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction