missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR109B/HM74 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24488SS
This item is not returnable.
View return policy
Description
GPR109B/HM74 Polyclonal specifically detects GPR109B/HM74 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| GPR109B/HM74 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide | |
| HCAR3 | |
| A portion of amino acids 1-50 of human GPR109B was used as the immunogen. | |
| 0.025 mg | |
| Apoptosis, Cancer, GPCR, Tumor Suppressors | |
| 8843 | |
| Human, Primate | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 0.5-2 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml | |
| G protein-coupled receptor 109B, G-protein coupled receptor 109B, G-protein coupled receptor HM74, G-protein coupled receptor HM74B, GTP-binding protein, HCA3, HCAR3, HM74B, HM74Puma-g, Niacin receptor 2, NIACR2, Nicotinic acid receptor 2, PUMAG, putative chemokine receptor | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 86% in rhesus monkey and 73% homologous in bovine and human REG3G. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction