missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR108 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24009-1-AP-20UL
This item is not returnable.
View return policy
Description
GPR108 is a protein coding gene.Specifications
| GPR108 | |
| Polyclonal | |
| Unconjugated | |
| GPR108 | |
| G protein coupled receptor 108, GPR108, LUSTR2, Protein GPR108 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 316136, 56927, 78308 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6P6V6, Q91WD0, Q9NPR9 | |
| Gpr108 | |
| GPR108 Fusion Protein Ag20887 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction