missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR108 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533603
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human (100%) Gorilla, Monkey, Marmoset, Pig (94%) Mouse, Rat, Bovine, Horse (89%) Panda (83%).
GPR108 is a protein coding gene.
Specifications
| GPR108 | |
| Polyclonal | |
| Unconjugated | |
| GPR108 | |
| 1810015L19Rik; AA589464; C79132; G protein-coupled receptor 108; GPR108; Lung seven transmembrane receptor 2; LUSTR2; Protein GPR108 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56927 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9NPR9 | |
| GPR108 | |
| Synthetic 18 amino acid peptide from 1st cytoplasmic domain of human GPR108. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction