missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR107 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102079
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GPR107.
GPR107 also named as KIAA1624 or LUSTR1 is 600 amino acid protein, which belongs to the LU7TM family. GPR107 localizes in membrane and is a multi-pass membrane protein. GPR107 is a promising candidate receptor for neuronostatin, and neuronostatin, interacting with GPR107, may play an important role in the central control of cardiovascular function.
Specifications
| GPR107 | |
| Polyclonal | |
| Unconjugated | |
| Gpr107 | |
| 9930033D15Rik; AI790205; bA138E2.2; C530034M11; G protein-coupled receptor 107; GCDRP; Gpr107; KIAA1624; Lung seven transmembrane receptor 1; LUSTR1; mKIAA1624; Protein GPR107 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 277463, 57720 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5VW38, Q8BUV8 | |
| Gpr107 | |
| A synthesized peptide derived from human GPR107(Accession Q5VW38), corresponding to amino acid residues R149-N199. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction