Learn More
Invitrogen™ GPR10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532993
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Rat (100%); Monkey, Marmoset, Mouse, Dog, Panda (95%); Bovine, Pig (89%); Horse (84%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
PRLHR is a 7-transmembrane domain receptor for prolactin-releasing hormone (PRLH) that is highly expressed in anterior pituitary (Ozawa et al., 2002).
Specifications
| GPR10 | |
| Polyclonal | |
| Unconjugated | |
| PRLHR | |
| G protein-coupled receptor 10; Gm339; GPR10; G-protein coupled receptor 10; GR3; hGR3; MGC126539; MGC126541; Prlhr; prolactin releasing hormone receptor; prolactin releasing peptide receptor; prolactin-releasing hormone receptor; prolactin-releasing peptide receptor; PrRP receptor; Prrpr; UHR-1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 246075, 2834 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P49683, Q64121 | |
| PRLHR | |
| Synthetic 19 amino acid peptide from 2nd extracellular domain of human PRLHR / GPR10. | |
| 50 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.