missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577691
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
PRLHR is a 7-transmembrane domain receptor for prolactin-releasing hormone (PRLH; MIM 602663) that is highly expressed in anterior pituitary (Ozawa et al., 2002 [PubMed 11923475]).
Specifications
| GPR10 | |
| Polyclonal | |
| Unconjugated | |
| Prlhr | |
| Prolactin-Releasing Peptide Receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 226278, 246075 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 0.85 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| Q64121, Q6VMN6 | |
| Prlhr | |
| Peptide (C)RVSVKLRNRVVPGS, corresponding to amino acid residues 247-260 of rat Prolactin-releasing peptide receptor | |
| 50 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction