missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587929
This item is not returnable.
View return policy
Description
Immunogen sequence: MASSTTRGPR VSDLFSGLPP AVTTPANQSA EASAGNGSVA GADAPAVTPF QSLQLVHQLK GLIVLLYSVV; Positive Samples: HepG2, U-87MG, Mouse brain; Cellular Location: Cell membrane, Multi-pass membrane protein.
PRLHR is a 7-transmembrane domain receptor for prolactin-releasing hormone (PRLH) that is highly expressed in anterior pituitary (Ozawa et al., 2002).
Specifications
| GPR10 | |
| Polyclonal | |
| Unconjugated | |
| PRLHR | |
| G protein-coupled receptor 10; Gm339; GPR10; G-protein coupled receptor 10; GR3; hGR3; MGC126539; MGC126541; Prlhr; prolactin releasing hormone receptor; prolactin releasing peptide receptor; prolactin-releasing hormone receptor; prolactin-releasing peptide receptor; PrRP receptor; Prrpr; UHR-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 226278, 2834 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P49683, Q6VMN6 | |
| PRLHR | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-70 of human PRLHR (NP_0042391). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction