missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPR10 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13404669
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13404669 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13404669 Supplier Invitrogen™ Supplier No. PA532994

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Dog, Bovine, Panda (100%); Monkey, Marmoset, Mouse, Rat, Pig (94%); Horse (89%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

PRLHR is a 7-transmembrane domain receptor for prolactin-releasing hormone (PRLH) that is highly expressed in anterior pituitary (Ozawa et al., 2002).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR10
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene PRLHR
Gene Accession No. P49683, Q4EW11
Gene Alias G protein-coupled receptor 10; Gm339; GPR10; G-protein coupled receptor 10; GR3; hGR3; MGC126539; MGC126541; Prlhr; prolactin releasing hormone receptor; prolactin releasing peptide receptor; prolactin-releasing hormone receptor; prolactin-releasing peptide receptor; PrRP receptor; Prrpr; UHR-1
Gene Symbols PRLHR
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from 3rd extracellular domain of human PRLHR / GPR10.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2834, 486911, 510397
Target Species Human, Canine, Bovine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.