Learn More
GPNMB Mouse anti-Human, CoraLite 594, Clone: 2B10B8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-66926-100UL
Description
GPNMB also known as HGFIN, osteoactivin, and DC-HIL, is a type I membrane glycoprotein involved in various biological processes, including inflammation, invasion and metastasis of malignant tumors, cell differentiation, and tissue regeneration. GPNMB shows expression in the lowly metastatic human melanoma cell lines and xenografts but does not show expression in the highly metastatic cell lines. GPNMB acts as an osteogenic factor that stimulates osteoblast differentiation in vivo and in vitro.
Specifications
| GPNMB | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| IgG1 |
| Immunofluorescence | |
| 2B10B8 | |
| CoraLite 594 | |
| 100 μL | |
| 10457 | |
| Liquid |