missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPN3 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18860PE
This item is not returnable.
View return policy
Description
GPN3 Polyclonal antibody specifically detects GPN3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| GPN3 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| ATP binding domain 1 family, member C, ATPBD1C, ATP-binding domain 1 family member C, GPN-loop GTPase 3, MGC14560, MGC32810, protein x 0004 | |
| Produced in rabbits immunized with E. coli-derived Human GPN3 fragment | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 51184 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction