missing translation for 'onlineSavingsMsg'
Learn More

GPN3 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-18860MFV450

Product Code. 18591259

  • £368.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

GPN3 Polyclonal antibody specifically detects GPN3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GPN3
Polyclonal
Immunohistochemistry-Paraffin
ATP binding domain 1 family, member C, ATPBD1C, ATP-binding domain 1 family member C, GPN-loop GTPase 3, MGC14560, MGC32810, protein x 0004
Produced in rabbits immunized with E. coli-derived Human GPN3 fragment
0.1 mL
Primary
Human
Purified
Immunohistochemistry (Paraffin)
mFluor Violet 450 SE
50mM Sodium Borate
Rabbit
Antigen and protein A Affinity-purified
RUO
51184
Store at 4C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.