missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588392
This item is not returnable.
View return policy
Description
Positive Samples: Mouse liver, Mouse lung, Mouse spleen; Cellular Location: Cytoplasm, Nucleus Immunogen sequence: VGVSAVLGTG LDELFVQVTS AAEEYEREYR PEYERLKKSL ANAESQQQRE QLERLRKDMG SVALDAGTAK DSLSPVLHPS DLILTRGTLD EEDEEADSDT DDIDHRVTEE SHEEPAFQNF.
Small GTPase required for proper nuclear import of RNA polymerase II (RNAPII) (PubMed:20855544, PubMed:21768307). May act at an RNAP assembly step prior to nuclear import (PubMed:21768307). Forms an interface between the RNA polymerase II enzyme and chaperone/scaffolding proteins, suggesting that it is required to connect RNA polymerase II to regulators of protein complex formation (PubMed:17643375). May be involved in nuclear localization of XPA (PubMed:11058119).
Specifications
| GPN1 | |
| Polyclonal | |
| Unconjugated | |
| Gpn1 | |
| 2410004J02Rik; AI449615; ATP(GTP)-binding protein; ATPBD1A; GPN1; GPN-loop GTPase 1; HUSSY-23; MBD2-interacting protein; MBDIN; methyl-CpG binding domain protein 2 interacting protein; NTPBP; RNA polymerase II associated protein 4; RNAPII-associated protein 4; RPAP4; XAB1; XPA binding protein 1; XPA binding protein 1, GTPase; XPA-binding protein 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11321, 74254 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8VCE2, Q9HCN4 | |
| Gpn1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 255-374 of human GPN1 (NP_009197.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction