missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPM6A Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £375.00
Specifications
| Antigen | GPM6A |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18330527
|
Novus Biologicals
NBP3-04744-20UL |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18342997
|
Novus Biologicals
NBP3-04744-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GPM6A Polyclonal specifically detects GPM6A in Mouse, Rat samples. It is validated for Western Blot.Specifications
| GPM6A | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 2823 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat | |
| glycoprotein M6A, M6a, M6AGPM6, neuronal membrane glycoprotein M6-a | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 159-278 of human GPM6A (NP_005268.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title