missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPM6A Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-03901-100UL
This item is not returnable.
View return policy
Description
GPM6A Polyclonal specifically detects GPM6A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| GPM6A | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 2823 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| glycoprotein M6A, M6a, M6AGPM6, neuronal membrane glycoprotein M6-a | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human GPM6A (NP_005268.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction