Learn More
GPKOW Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14728-1-AP-20UL
 View more versions of this product
Product Code. 16803524
  • £135.00 / 20µL
Description
This gene encodes a putative RNA-binding protein containing G-patch and KOW (Kyprides, Ouzounis, Woese) domains. The encoded protein interacts directly with protein kinase A and protein kinase X and is also found associated with the spliceosome.
Specifications
Specifications
GPKOW
Polyclonal
Unconjugated
GPKOW
G patch domain and KOW motifs, GPATC5, GPATCH5, GPKOW, Protein MOS2 homolog, Protein T54, T54
Rabbit
Antigen Affinity Chromatography
RUO
209416, 27238, 679890
-20°C
Liquid
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q56A08, Q92917
GPKOW
GPKOW Fusion Protein Ag6487
20 μL
Primary
Human, Rat, Mouse