missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPKOW Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531000
This item is not returnable.
View return policy
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a putative RNA-binding protein containing G-patch and KOW (Kyprides, Ouzounis, Woese) domains. The encoded protein interacts directly with protein kinase A and protein kinase X and is also found associated with the spliceosome.
Specifications
| GPKOW | |
| Polyclonal | |
| Unconjugated | |
| GPKOW | |
| 4432411B18; DXErtd697e; DXImx41e; G patch domain and KOW motifs; G patch domain and KOW motifs-containing protein; GPATC5; G-patch domain and KOW motifs; G-patch domain and KOW motifs-containing protein; G-patch domain-containing protein 5; GPATCH5; GPKOW; Protein MOS2 homolog; Protein T54; Spp2; T54 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27238 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q92917 | |
| GPKOW | |
| Recombinant protein encompassing a sequence within the center region of human GPKOW. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction