Learn More
Invitrogen™ GPKOW Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589085
Beschreibung
Immunogen sequence: TLAIPMIQKG CTPSGEGADS EPRAETVPEE ANYEAVPVEA YGLAMLRGMG WKPGEGIGRT FNQVVKPRVN SLRPKGLGLG ANLTEAQALT PTGPSRMPRP DEEQEKDKED QPQGLVPGGA VVVLSGPHRG LYGKVEGLDP DNVRAMVRLA VGSRVVTVSE YYLRPVSQQE FDKNTLDLRQ QNGTASSRKT LWNQELYIQQ DNSERKRKHL PDRQDGPAAK SEKAAPRSQH WLHRDLRVRF VDNMYKGGQY YNTKMIIEDV LSPDTCVCRT DEGRVLEGLR EDMLETLVPK AEGDRVMVVL GPQTGRVGHL LSRDRARSRA LVQLPRENQV VELHYDAICQ YMGPSDTDDD; Positive Samples: Mouse brain, Mouse kidney, Rat brain; Cellular Location: Nucleus.
This gene encodes a putative RNA-binding protein containing G-patch and KOW (Kyprides, Ouzounis, Woese) domains. The encoded protein interacts directly with protein kinase A and protein kinase X and is also found associated with the spliceosome.
Spezifikation
| GPKOW | |
| Polyclonal | |
| Unconjugated | |
| GPKOW | |
| 4432411B18; DXErtd697e; DXImx41e; G patch domain and KOW motifs; G patch domain and KOW motifs-containing protein; GPATC5; G-patch domain and KOW motifs; G-patch domain and KOW motifs-containing protein; G-patch domain-containing protein 5; GPATCH5; GPKOW; Protein MOS2 homolog; Protein T54; Spp2; T54 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 209416, 27238, 679890 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.61 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q56A08, Q92917 | |
| GPKOW | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 127-476 of human GPKOW (NP_0565132). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.