missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPIP137 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15721671
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15721671 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15721671 Supplier Invitrogen™ Supplier No. PA553733

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LWDLLEGKEK PVCGTTYKVL KEIVERVFQS NYFDSTHNHQ NGLCEEEEAA SAPAVEDQVP EAEPEPAEEY TEQSEVESTE YVNRQFMAET QFTSGEKEQV DEWTVETVEV VNSLQQQPQA ASPSVPEPHS LTPVAQ Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.

GPIP137 regulates the transport and translation of mRNAs of proteins involved in synaptic plasticity in neurons and cell proliferation/migration in multiple cell types. It binds directly and selectively to MYC and CCND2 RNAs. In neuronal cells, GPIP137 binds to several mRNAs associated with RNA granules including BDNF, CAMK2A, CREB1, MAP2, NTRK2, mRNAs, GRIN1, and KPNB1 mRNAs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPIP137
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Caprin1
Gene Accession No. Q14444
Gene Alias activation/proliferation-associated protein 1; AL022980; caprin 1; Caprin1; caprin-1; cell cycle associated protein 1; cell cycle-associated protein 1; cytop; Cytoplasmic activation- and proliferation-associated protein 1; cytoplasmic activation/proliferation-associated protein 1; cytoplasmic activation/proliferation-associated protein-1; G5E5; GPI p137; GPI-anchored membrane protein 1; GPI-anchored protein p137; Gpiap; GPIAP1; Gpip137; GPI-p137; GRIP137; M11S1; Membrane component chromosome 11 surface marker 1; membrane component, chromosome 11, surface marker 1; Mmgpip137; P137; p137GPI; RNA granule protein 105; Rng105
Gene Symbols Caprin1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPIP137. Recombinant protein control fragment (Product #RP-90904).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4076
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.