missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ GPIP137 Antibody (Toray patent anti-Caprin-1) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28615-1mg
This item is not returnable.
View return policy
Description
GPIP137 Monoclonal antibody specifically detects GPIP137 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| GPIP137 | |
| Monoclonal | |
| Unconjugated | |
| Q14444 | |
| Human | |
| Protein A purified | |
| RUO | |
| 4076 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| Toray patent anti-Caprin-1 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| activation/proliferation-associated protein 1, caprin 1, caprin-1, cell cycle associated protein 1, Cell cycle-associated protein 1, Cytoplasmic activation- and proliferation-associated protein 1, cytoplasmic activation/proliferation-associated protein-1, GPI-anchored membrane protein 1GPIP137, GPI-p137, M11S1GPI-anchored protein p137, Membrane component chromosome 11 surface marker 1, membrane component, chromosome 11, surface marker 1, p137GPI, RNA granule protein 105, RNG105GPIAP1 | |
| CAPRIN1 | |
| 1 mg | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction