missing translation for 'onlineSavingsMsg'
Learn More

GPIP137 Antibody, HRP, Novus Biologicals™

Product Code. 18415871 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18415871 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18415871 Supplier Novus Biologicals Supplier No. NBP222238H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GPIP137 Polyclonal antibody specifically detects GPIP137 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPIP137
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot
Formulation PBS
Gene Alias activation/proliferation-associated protein 1, caprin 1, caprin-1, cell cycle associated protein 1, Cell cycle-associated protein 1, Cytoplasmic activation- and proliferation-associated protein 1, cytoplasmic activation/proliferation-associated protein-1, GPI-anchored membrane protein 1GPIP137, GPI-p137, M11S1GPI-anchored protein p137, Membrane component chromosome 11 surface marker 1, membrane component, chromosome 11, surface marker 1, p137GPI, RNA granule protein 105, RNG105GPIAP1
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 550 and 600 of human Cell Cycle Associated Protein 1 using the numbering given in entry NP_005889.3 (GeneID 4076).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4076
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.