missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPIHBP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA116995
This item is not returnable.
View return policy
Description
GPIHBP1 Polyclonal Antibody for Western Blot
GPI-anchored HDL-binding protein 1 (Gpihbp1) is a novel protein that may be involved in the regulation of the delivery of fats to cells for energy and storage. Digested fats travel to the small intestine, where they are packaged into chylomicrons (particles filled with triglycerides). Chylomicrons then travel through the bloodstream and deliver triglycerides to tissues that are hungry for fuel or to adipose tissue for energy storage. Triglycerides are broken down or hydrolyzed by the enzyme lipoprotein lipase (LpL). The triglyceride breakdown products are then taken up and used by cells. Gpihbp1 is the molecule in capillaries that facilitates the capture of chylomicrons and facilitates the interaction with LpL. It has been shown that fats in the bloodstream are not readily metabolized in the absence of GPIHBP1.
Specifications
| GPIHBP1 | |
| Polyclonal | |
| Unconjugated | |
| GPIHBP1 | |
| 1110002J19Rik; endothelial cell LPL transporter; glycosylphosphatidylinositol anchored high density lipoprotein binding protein 1; glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein 1; GPI anchored high density lipoprotein binding protein 1; GPI-anchored HDL-binding protein 1; Gpihbp1; GPI-HBP1; Hbp1; HDLBP; HDLBP1; High density lipoprotein-binding protein 1; HYPL1D; RGD1564237 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 338328 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| TBS with 0.05% sodium azide | |
| Q8IV16 | |
| GPIHBP1 | |
| A synthetic peptide corresponding to an internal region [within residues 100-180] of human GPIHBP1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction