missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPIHBP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£188.00 - £417.00
Specifications
| Antigen | GPIHBP1 |
|---|---|
| Concentration | 0.9 mg/mL |
| Dilution | Western Blot 2.0 ug/ml, Immunocytochemistry/Immunofluorescence 1:20 - 1:50 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18139699
|
Novus Biologicals
NB110-41539SS |
0.025 mL |
£188.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18726243
|
Novus Biologicals
NB110-41539 |
0.1 mL |
£417.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GPIHBP1 Polyclonal antibody specifically detects GPIHBP1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.Specifications
| GPIHBP1 | |
| Western Blot 2.0 ug/ml, Immunocytochemistry/Immunofluorescence 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism, Signal Transduction | |
| Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide | |
| 338328 | |
| A synthetic peptide corresponding to an internal region [within residues 170-250] of mouse GPIHBP1. [Swiss-Prot# Q9D1N2] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| 0.9 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Mouse | |
| glycosylphosphatidylinositol anchored high density lipoprotein binding protein1, glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein1, GPI anchored high density lipoprotein binding protein 1, GPI-anchored HDL-binding protein 1, GPI-HBP1LOC338328, HBP1, High density lipoprotein-binding protein 1 | |
| GPIHBP1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title