missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPIHBP1 Antibody (1F9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00338328-M02
This item is not returnable.
View return policy
Description
GPIHBP1 Monoclonal antibody specifically detects GPIHBP1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| GPIHBP1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| glycosylphosphatidylinositol anchored high density lipoprotein binding protein1, glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein1, GPI anchored high density lipoprotein binding protein 1, GPI-anchored HDL-binding protein 1, GPI-HBP1LOC338328, HBP1, High density lipoprotein-binding protein 1 | |
| GPIHBP1 (NP_835466.1, 1 a.a. ∽ 184 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MKALGAVLLALLLCGRPGRGQTQQEEEEEDEDHGPDDYDEEDEDEVEEEETNRLPGGRSRVLLRCYTCKSLPRDERCNLTQNCSHGQTCTTLIAHGNTESGLLTTHSTWCTDSCQPITKTVEGTQVTMTCCQSSLCNVPPWQSSRVQDPTGKGAGGPRGSSETVGAALLLNLLAGLGAMGARRP | |
| 0.1 mg | |
| Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
| 338328 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA | |
| 1F9 | |
| Western Blot, ELISA 0.03 ng/mL | |
| NP_835466.1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction