missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPI Recombinant Rabbit Monoclonal Antibody (23GB5595), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553906
This item is not returnable.
View return policy
Description
This gene belongs to the GPI family whose members encode multifunctional phosphoglucose isomerase proteins involved in energy pathways. The protein encoded by this gene is a dimeric enzyme that catalyzes the reversible isomerization of glucose-6-phosphate and fructose-6-phosphate. The protein functions in different capacities inside and outside the cell. In the cytoplasm, the gene product is involved in glycolysis and gluconeogenesis, while outside the cell it functions as a neurotrophic factor for spinal and sensory neurons. Defects in this gene are the cause of nonspherocytic hemolytic anemia and a severe enzyme deficiency can be associated with hydrops fetalis, immediate neonatal death and neurological impairment.
Specifications
| GPI | |
| Recombinant Monoclonal | |
| Unconjugated | |
| GPI | |
| Amf; Autocrine motility factor; glucose phosphate isomerase; glucose phosphate isomerase 1; glucose phosphate isomerase 1 complex; glucose phosphate isomerase 1, regulatory; glucose phosphate isomerase 1, structural; glucose phosphate isomerase 1, temporal; glucose-6-phosphate isomerase; glucose-6-phosphate isomerase {ECO:0000250; GNPI; Gpi; GPI {ECO:0000250; gpi {ECO:0000312; GPI1; Gpi-1; Gpi-1r; Gpi1-r; Gpi1s; Gpi-1s; Gpi1-s; Gpi-1t; Gpi1-t; GPI-GlcNAc transferase complex subunit GPI1; hexose monophosphate isomerase; hexosephosphate isomerase; maturation factor; MF; Neuroleukin; neuroleukin {ECO:0000250; NK; NK/GPI; Nlk; NLK {ECO:0000250; Org; oxoisomerase; PGI; PGI {ECO:0000250; Phi; PHI {ECO:0000250; phosphatidylinositol N-acetylglucosaminyltransferase; Phosphatidylinositol N-acetylglucosaminyltransferase subunit GPI1; Phosphoglucose isomerase; phosphoglucose isomerase {ECO:0000250; phosphohexomutase; phosphohexose isomerase; phosphohexose isomerase {ECO:0000250; phosphosaccharomutase; RGD:2727}; SA36; SA-36; sperm antigen; sperm antigen 36; sperm antigen-36; UniProtKB:P06745}; YGR216C | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 2821 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5595 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P06744 | |
| GPI | |
| A synthesized peptide derived from human Glucose 6 phosphate isomerase (1-64AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction