Learn More
Invitrogen™ GPI Monoclonal Antibody (K94005_19C2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555714
Description
Sequence of this protein is as follows: MAALTRDPQF QKLQQWYREH RSELNLRRLF DANKDRFNHF SLTLNTNHGH ILVDYSKNLV TEDVMRMLVD LAKSRGVEAA RERMFNGEKI NYTEGRAVLH VALRNRSNTP ILVDGKDVMP EVNKVLDKMK SFCQRVRSGD WKGYTGKTIT DVINIGIGGS DLGPLMVTEA LKPYSSGGPR VWYVSNIDGT HIAKTLAQLN PESSLFIIAS KTFTTQETIT NAETAKEWFL QAAKDPSAVA KHFVALSTNT TKVKEFGIDP QNMFEFWDWV GGRYSLWSAI GLSIALHVGF DNFEQLLSGA HWMDQHFRTT PLEKNAPVLL ALLGIWYINC FGCETHAMLP YDQYLHRFAA YFQQGDMESN GKYITKSGTR VDHQTGPIVW GEPGTNGQHA FYQLIHQGTK MIPCDFLIPV QTQHPIRKGL HHKILLANFL AQTEALMRGK STEEARKELQ AAGKSPEDLE RLLPHKVFEG NRPTNSIVFT KLTPFMLGAL VAMYEHKIFV QGIIWDINSF DQWGVELGKQ LAKKIEPELD GSAQVTSHDA STNGLINFIK QQREARVQ
This gene belongs to the GPI family whose members encode multifunctional phosphoglucose isomerase proteins involved in energy pathways. The protein encoded by this gene is a dimeric enzyme that catalyzes the reversible isomerization of glucose-6-phosphate and fructose-6-phosphate. The protein functions in different capacities inside and outside the cell. In the cytoplasm, the gene product is involved in glycolysis and gluconeogenesis, while outside the cell it functions as a neurotrophic factor for spinal and sensory neurons. Defects in this gene are the cause of nonspherocytic hemolytic anemia and a severe enzyme deficiency can be associated with hydrops fetalis, immediate neonatal death and neurological impairment.
Specifications
| GPI | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P06744 | |
| GPI | |
| Recombinant human GPI (immunogen range is 1-558). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunoprecipitation, Western Blot | |
| K94005_19C2 | |
| Unconjugated | |
| GPI | |
| Amf; Autocrine motility factor; glucose phosphate isomerase; glucose phosphate isomerase 1; glucose phosphate isomerase 1 complex; glucose phosphate isomerase 1, regulatory; glucose phosphate isomerase 1, structural; glucose phosphate isomerase 1, temporal; glucose-6-phosphate isomerase; glucose-6-phosphate isomerase {ECO:0000250; GNPI; Gpi; GPI {ECO:0000250; gpi {ECO:0000312; GPI1; Gpi-1; Gpi-1r; Gpi1-r; Gpi1s; Gpi-1s; Gpi1-s; Gpi-1t; Gpi1-t; GPI-GlcNAc transferase complex subunit GPI1; hexose monophosphate isomerase; hexosephosphate isomerase; maturation factor; MF; Neuroleukin; neuroleukin {ECO:0000250; NK; NK/GPI; Nlk; NLK {ECO:0000250; Org; oxoisomerase; PGI; PGI {ECO:0000250; Phi; PHI {ECO:0000250; phosphatidylinositol N-acetylglucosaminyltransferase; Phosphatidylinositol N-acetylglucosaminyltransferase subunit GPI1; Phosphoglucose isomerase; phosphoglucose isomerase {ECO:0000250; phosphohexomutase; phosphohexose isomerase; phosphohexose isomerase {ECO:0000250; phosphosaccharomutase; RGD:2727}; SA36; SA-36; sperm antigen; sperm antigen 36; sperm antigen-36; UniProtKB:P06745}; YGR216C | |
| Mouse | |
| Protein A | |
| RUO | |
| 2821 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.