missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPD2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99207-50ul
This item is not returnable.
View return policy
Description
GPD2 Polyclonal antibody specifically detects GPD2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| GPD2 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:1000-1:4000 | |
| EC 1.1.5.3, GDH2, glycerol-3-phosphate dehydrogenase 2 (mitochondrial), GPDM, GPD-M, mGPDH, mitochondrial, mitochondrial glycerophosphate dehydrogenase, mtGPD | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human GPD2. | |
| 50 μg | |
| Lipid and Metabolism | |
| 2820 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction