missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPD1L Mouse anti-Human, Clone: 1.40E+03, Novus Biologicals™
Mouse Monoclonal Antibody
£234.00 - £381.00
Specifications
| Antigen | GPD1L |
|---|---|
| Clone | 1400 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500, Flow Cytometry 1:10-1:1045, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18287101
|
Novus Biologicals
NBP2-59439-50UL |
50 μL |
£234.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18242661
|
Novus Biologicals
NBP2-59439-100UL |
100 μL |
£381.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GPD1L Monoclonal specifically detects GPD1L in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| GPD1L | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| EC 1.1.1, EC 1.1.1.8, glycerol-3-phosphate dehydrogenase 1-like, glycerol-3-phosphate dehydrogenase 1-like protein, KIAA0089GPD1-L | |
| GPD1L | |
| IgG1 κ | |
| Protein A purified |
| 1400 | |
| Western Blot 1:500, Flow Cytometry 1:10-1:1045, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| 23171 | |
| Recombinant human GPD1L (1-351aa) purified from E. coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title