missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPCR GPR89 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | GPCR GPR89 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18295543
|
Novus Biologicals
NBP2-55925 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18611739
|
Novus Biologicals
NBP2-55925-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GPCR GPR89 Polyclonal specifically detects GPCR GPR89 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| GPCR GPR89 | |
| Polyclonal | |
| Rabbit | |
| GPCR | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 653519 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LGDPFPILSPKHGILSIEQL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| FLJ10492, FLJ11696, FLJ36909, FLJ79270, G protein-coupled receptor 89A, Golgi pH regulator A, GPHRA, GPR89, GPR89B, MGC688, MGC87324, protein GPR89, Protein GPR89A, Putative MAPK-activating protein PM01, Putative NF-kappa-B-activating protein 90, SH120, UNQ192 | |
| GPR89A | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title