missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPCR/G-protein complex-stabilizing scFv Recombinant Mouse Monoclonal Antibody (scFv16)

Mouse Recombinant Monoclonal Antibody

£456.00 - £1842.00

Specifications

Antigen GPCR/G-protein complex-stabilizing scFv
Clone scFv16
Concentration 2.0 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications Not Available
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17184679
View Documents
Invitrogen™
703976
100 μg
£456.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17194679
View Documents
Invitrogen™
7039761MG
1 mg
£1842.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The scFv antibody stabilizes GPCR/G-protein complexes for cryo-EM of active structures. It recognizes an interface between G alpha and G beta/gamma subunits, and confers resistance to nucleotide exchange-triggered dissociation of the heterotrimeric G-protein. In addition to members of the Gi/o family, this antibody fragment can also be applied to chimeric variants and stabilize other G-protein subtypes including Gs complexes.

Specifications

Specifications

GPCR/G-protein complex-stabilizing scFv
2.0 mg/mL
Not Available
Unconjugated
Mouse
Human
Rhodopsin/Gi1.
Antibody
scFv16
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Liquid
RUO
25mM HEPES with 10% glycerol and no preservative; pH 7.5
Primary
Affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.