Learn More
GPBB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532782
Code : PN
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Orangutan, Gibbon, Marmoset, Mouse, Rat, Sheep, Bat, Bovine, Hamster, Elephant, Panda, Rabbit, Pig, Turkey, Chicken, Lizard, Xenopus, Salmon, Pufferfish, Zebrafish, Stickleback, Drosophila (100%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
The protein encoded by this gene is a glycogen phosphorylase found predominantly in the brain. The encoded protein forms homodimers which can associate into homotetramers, the enzymatically active form of glycogen phosphorylase. The activity of this enzyme is positively regulated by AMP and negatively regulated by ATP, ADP, and glucose-6-phosphate. This enzyme catalyzes the rate-determining step in glycogen degradation.Specifications
GPBB | |
Polyclonal | |
Unconjugated | |
PYGB | |
brain glycogen phosphorylase; glycogen phosphorylase B; glycogen phosphorylase, brain form; GLYPHOA; GPBB; MGC9213; phosphorylase, glycogen; brain; PYGB | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100342407, 100621536, 100769186, 110078, 25739, 421248, 505560, 554325, 5834 | |
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P11216, P53534, Q3B7M9, Q5MIB6, Q8CI94 | |
PYGB | |
Synthetic 20 amino acid peptide from internal region of human PYGB. | |
50 μg | |
Primary | |
Bovine, Chicken, Drosophila, Chicken, Human, Hamster, Human, Mouse, Non-human Primate, Ovine, Porcine, Rabbit | |
Antibody | |
IgG |