missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPATCH4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24399-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is a rabbit polyclonal antibody raised against the 189 residues of N-terminal of human GPATCH4 protein.
The function of this protein remains unknown.Specifications
| GPATCH4 | |
| Polyclonal | |
| Unconjugated | |
| GPATCH4 | |
| G patch domain containing 4, GPATC4, GPATCH4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 295228, 54865, 66614 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q3TFK5, Q566R3, Q5T3I0 | |
| Gpatch4 | |
| GPATCH4 Fusion Protein Ag20095 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction